(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98.0%
Supplier: TCI America
[for Determination of the optical purity of Alcohols and Amines]
CAS Number: 20445-33-4
MDL Number: MFCD00067105
Molecular Formula: C10H8ClF3O2
Molecular Weight: 252.62
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): 89
Specific Gravity (20/20): 1.35
Specific rotation [a]20/D: 149 deg (C=2, Toluene)
Product Categories: Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (+)-Mosher's Acid Chloride, (+)-MTPA-Cl
Formula:
C₁₀H₈ClF₃O₂ Boiling Pt: 99 °C Density: 1.35 (20 °C) Flash Pt: 89 °C |
MDL Number:
MFCD00067105 CAS Number: 20445-33-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...