Methacroylcholine chloride ∼80% in water stabilised

Supplier: TCI America

Synonyms: 2-(Methacryloyloxy)-N,N,N-trimethylethanaminium chloride, (Methacryloyloxyethyl)trimethyl ammonium chloride, 2-(Methacryloyloxy)ethyltrimethylammonium chloride, Trimethyl-2-methacroyloxyethylammonium Chloride

M0918-25G M0918-500G
TCM0918-025GEA 17.3 USD
TCM0918-025G TCM0918-500G
Methacroylcholine chloride ∼80% in water stabilised
Methacroylcholine chloride

(stabilized with MEHQ)
CAS Number: 5039-78-1
MDL Number: MFCD00060097
Molecular Formula: C9H18ClNO2
Molecular Weight: 207.70
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: Trimethyl-2-methacroyloxyethylammonium Chloride

Formula: C₉H₁₈ClNO₂
Density: 1.11 (20 °C)
MDL Number: MFCD00060097
CAS Number: 5039-78-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09b82a06af4540088146e71d2df2e6b1f3e1ba5a&vid=992d390a9c2d27fd0df314d3ec6f0005600dfa78&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR