N-Methylhydroxylammonium chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Methylhydroxylamine hydrochloride

M0804-25G M0804-5G
TCM0804-025GEA 142 USD
TCM0804-025G TCM0804-005G
N-Methylhydroxylammonium chloride ≥97.0% (by titrimetric analysis)
N-Methylhydroxylammonium chloride

CAS Number: 4229-44-1
MDL Number: MFCD00012597
Molecular Formula: CH5NO
Molecular Weight: 83.52
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 86

Formula: CH₆ClNO
MW: 83.52 g/mol
Melting Pt: 86 °C
MDL Number: MFCD00012597
CAS Number: 4229-44-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f580eb504886cfe62387e938bbcef2ff03919bcc&vid=f580eb504886cfe62387e938bbcef2ff03919bcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR