3-Methyl-2-butenal ≥97.0%

Supplier: TCI America

Synonyms: 3,3-Dimethylacrolein, 3-Methylbut-2-enal, Senecialdehyde, 3-Methylcrotonaldehyde

M0770-25ML M0770-100ML
TCM0770-025MLEA 151 USD
TCM0770-025ML TCM0770-100ML
3-Methyl-2-butenal ≥97.0%
3-Methyl-2-butenal

CAS Number: 107-86-8
MDL Number: MFCD00010291
Molecular Formula: C5H8O
Molecular Weight: 84.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Melting point (°C): -20
Flash Point (°C): 37
Specific Gravity (20/20): 0.88
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3,3-Dimethylacrolein, 3-Methylcrotonaldehyde, Senecialdehyde

Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 135 °C
Melting Pt: –20 °C
Density: 0.88 (20 °C)
Flash Pt: 37 °C
MDL Number: MFCD00010291
CAS Number: 107-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d842a0bcd69e67b2a6d4f4962e45cbadb28c122e&vid=d842a0bcd69e67b2a6d4f4962e45cbadb28c122e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR