Vinyl methacrylate ≥98.0% stabilised
Supplier: TCI America
Synonyms:
Methacrylic Acid Vinyl Ester
M0720-500ML
M0720-25ML
M0720-100ML
TCM0720-500MLEA
410
USD
TCM0720-500ML
TCM0720-025ML
TCM0720-100ML
Vinyl methacrylate ≥98.0% stabilised
Vinyl methacrylate
(stabilized with MEHQ)
CAS Number: 4245-37-8
MDL Number: MFCD00059329
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 16
Specific Gravity (20/20): 0.94
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid Vinyl Ester
Formula:
C₆H₈O₂ MW: 112.13 g/mol Boiling Pt: 112 °C Density: 0.94 (20 °C) Flash Pt: 16 °C |
MDL Number:
MFCD00059329 CAS Number: 4245-37-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...