4,4'-Methylenebis(cyclohexylamine) (mixture of isomers) ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4,4'-Diaminodicyclohexylmethane, 4,4-Diaminodicyclohexyl methane, 4,4'-Methylenebis(aminocyclohexane), Bis(4-aminocyclohexyl)methane

M0699-500G M0699-25G
TCM0699-500GEA 126.6 USD
TCM0699-500G TCM0699-25G
4,4'-Methylenebis(cyclohexylamine) (mixture of isomers) ≥97.0% (by GC, titration analysis)
4,4'-Methylenebis(cyclohexylamine) (mixture of isomers)

CAS Number: 1761-71-3
MDL Number: MFCD00001496
Molecular Formula: C13H26N2
Molecular Weight: 210.37
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 126
Flash Point (°C): 162
Freezing point (°C): 40


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: Bis(4-aminocyclohexyl)methane, 4,4'-Diaminodicyclohexylmethane, 4,4'-Methylenebis(aminocyclohexane)

Formula: C₁₃H₂₆N₂
MW: 210.37 g/mol
Boiling Pt: 126 °C
Flash Pt: 162 °C
MDL Number: MFCD00001496
CAS Number: 1761-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cedcb55d3a5de6b7e2ccbd638d39a1ea94fd3619&vid=cedcb55d3a5de6b7e2ccbd638d39a1ea94fd3619&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR