Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers) ≥80.0%

Supplier: TCI America

Synonyms: Methylnadic Anhydride, Methylnadic anhydride, Nadic Methyl Anhydride, Methyl-3,6-endomethylenetetrahydrophthalic Anhydride

M0690-500G M0690-25G
TCM0690-500GEA 76 USD
TCM0690-500G TCM0690-25G
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers) ≥80.0%
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers)

CAS Number: 25134-21-8
MDL Number: MFCD00167036
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Flash Point (°C): 135
Specific Gravity (20/20): 1.23


Other Product Names / Synonyms: Methyl-3,6-endomethylenetetrahydrophthalic Anhydride, Methylnadic Anhydride

Formula: C₁₀H₁₀O₃
Density: 1.23 (20 °C)
Flash Pt: 135 °C
MDL Number: MFCD00167036
CAS Number: 25134-21-8

Order Now

Specification Test Results

Purity/Analysis Method >80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR