N-Methyl-1,3-propanediamine ≥98.0%
Supplier: TCI America
Synonyms:
N-Methylpropane-1,3-diamine, 3-Methylamino propylamine
M0599-25ML
M0599-100ML
TCM0599-25MLEA
59
USD
TCM0599-25ML
TCM0599-100ML
N-Methyl-1,3-propanediamine ≥98.0%
N-Methyl-1,3-propanediamine
CAS Number: 6291-84-5
MDL Number: MFCD00008209
Molecular Formula: C4H12N2
Molecular Weight: 88.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 143
Flash Point (°C): 36
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: 3-Methylaminopropylamine, N-Methyl-1,3-propanediamine
Formula:
C₄H₁₂N₂ MW: 88.15 g/mol Boiling Pt: 143 °C Density: 0.86 (20 °C) Flash Pt: 36 °C |
MDL Number:
MFCD00008209 CAS Number: 6291-84-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...