1-Methyl-1,4-cyclohexadiene ≥95.0%
Supplier: TCI America
M0486-5ML
M0486-25ML
TCM0486-005MLEA
240.01
USD
TCM0486-005ML
TCM0486-025ML
1-Methyl-1,4-cyclohexadiene ≥95.0%
1-Methyl-1,4-cyclohexadiene
CAS Number: 4313-57-9
MDL Number: MFCD00001538
Molecular Formula: C7H10
Molecular Weight: 94.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 11
Specific Gravity (20/20): 0.85
Other Product Names / Synonyms: 2,5-Dihydrotoluene
Formula:
C₇H₁₀ MW: 94.16 g/mol Boiling Pt: 115 °C Density: 0.85 (20 °C) Flash Pt: 11 °C |
MDL Number:
MFCD00001538 CAS Number: 4313-57-9 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...