Mucobromic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

M0471-500G M0471-25G
TCM0471-500GEA 766.8 USD
TCM0471-500G TCM0471-25G
Mucobromic acid ≥99.0% (by titrimetric analysis)
Mucobromic acid

CAS Number: 488-11-9
MDL Number: MFCD00063745
Molecular Formula: C4H2Br2O3
Molecular Weight: 257.87
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 124

Formula: C₄H₂Br₂O₃
Melting Pt: 124 °C
MDL Number: MFCD00063745
CAS Number: 488-11-9
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1913b25c64325a6577ef51945889fdc167bc80ea&vid=1913b25c64325a6577ef51945889fdc167bc80ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR