Metaldehyde ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Metacetaldehyde, Aldehyde (meta), Acetaldehyde Tetramer
M0076-500G
M0076-25G
TCM0076-500GEA
102
USD
TCM0076-500G
TCM0076-25G
Metaldehyde ≥97.0% (by titrimetric analysis)
Metaldehyde
CAS Number: 108-62-3
MDL Number: MFCD00134115
Molecular Formula: C8H16O4
Molecular Weight: 176.21
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 65
Melting point (°C): 246
Flash Point (°C): 50
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Aldehyde (meta), Metacetaldehyde, Acetaldehyde Tetramer
Formula:
C₈H₁₆O₄ MW: 176.21 g/mol Boiling Pt: 65 °C Melting Pt: 246 °C Flash Pt: 50 °C |
MDL Number:
MFCD00134115 CAS Number: 108-62-3 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...