Methacholine bromide ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: O-Acetyl-beta-methylcholine Bromide, Acetyl-β-methylcholine bromide, Mecholyl Bromide

M0072-25G M0072-100G
TCM0072-025GEA 99.4 USD
TCM0072-025G TCM0072-100G
Methacholine bromide ≥99.0% (by titrimetric analysis)
Methacholine bromide

CAS Number: 333-31-3
MDL Number: MFCD00011816
Molecular Formula: C8H18BrNO2
Molecular Weight: 240.14
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 147


Other Product Names / Synonyms: O-Acetyl-beta-methylcholine Bromide, Mecholyl Bromide

Formula: C₈H₁₈BrNO₂
MW: 240.14 g/mol
Melting Pt: 147 °C
MDL Number: MFCD00011816
CAS Number: 333-31-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26d0052bd8465690c1e18139cbefaf3d46dd7ee4&vid=26d0052bd8465690c1e18139cbefaf3d46dd7ee4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR