L-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: H-Lys-OMe·2HCl, H-Lys-OMe.2HCl

L0202-25G L0202-5G
TCL0202-25GEA 109 USD
TCL0202-25G TCL0202-5G
L-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen basis)
L-Lysine methyl ester dihydrochloride

CAS Number: 26348-70-9
MDL Number: MFCD00039067
Molecular Formula: C7H16N2O2
Molecular Weight: 233.13
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 197
Specific rotation [a]20/D: 17 deg (C=5, H2O)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Lys-OMe.2HCl

Formula: C₇H₁₆N₂O₂
MW: 233.13 g/mol
Melting Pt: 197 °C
MDL Number: MFCD00039067
CAS Number: 26348-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e72dc62c383d2a0615c01e9e095ad5b84b4ee10&vid=b4245de7f82aa1388a908ed596a90362e45201ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR