L-Leucine ethyl ester hydrochloride ≥99.0%
Supplier: TCI America
Synonyms:
H-Leu-OEt.HCl
L0123-25G
L0123-5G
TCL0123-025GEA
62
USD
TCL0123-025G
TCL0123-5G
L-Leucine ethyl ester hydrochloride ≥99.0%
L-Leucine ethyl ester hydrochloride
CAS Number: 2743-40-0
MDL Number: MFCD00034879
Molecular Formula: C8H17NO2
Molecular Weight: 195.69
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 136
Specific rotation [a]20/D: 19 deg (C=5, EtOH)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Leu-OEt.HCl
Formula:
C₈H₁₇NO₂ MW: 195.69 g/mol Melting Pt: 136 °C |
MDL Number:
MFCD00034879 CAS Number: 2743-40-0 EINECS: 220-375-6 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 99.0 % |
Specific rotation [a]20/D | +18.0 to +20.0 deg (C=5, EtOH) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...