2'-Iodoacetophenone ≥98.0%
Supplier: TCI America
Synonyms:
1-(2-Iodophenyl)ethan-1-one, 1-(2-Iodophenyl)ethanone, o-Iodoacetophenone
I0689-5G
I0689-1G
TCI0689-5GEA
47
USD
TCI0689-5G
TCI0689-1G
2'-Iodoacetophenone ≥98.0%
2'-Iodoacetophenone
CAS Number: 2142-70-3
MDL Number: MFCD00094998
Molecular Formula: C8H7IO
Molecular Weight: 246.05
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 103
Specific Gravity (20/20): 1.77
Formula:
C₈H₇IO MW: 246.05 g/mol Boiling Pt: 144 °C Density: 1.77 (20 °C) Flash Pt: 103 °C |
MDL Number:
MFCD00094998 CAS Number: 2142-70-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...