Methyl-4-iodo-3-methylbenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 4-iodo-3-methylbenzoate

I0601-25G I0601-5G
TCI0601-25GEA 289 USD
TCI0601-25G TCI0601-5G
Methyl-4-iodo-3-methylbenzoate ≥98.0%
Methyl 4-iodo-3-methylbenzoate

CAS Number: 5471-81-8
MDL Number: MFCD02683863
Molecular Formula: C9H9IO2
Molecular Weight: 276.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 106
Melting point (°C): 61


Other Product Names / Synonyms: 4-Iodo-m-toluic Acid Methyl Ester, 4-Iodo-3-methylbenzoic Acid Methyl Ester, Methyl 4-Iodo-m-toluate

Formula: C₉H₉IO₂
MW: 276.07 g/mol
Boiling Pt: 106 °C
Melting Pt: 61 °C
MDL Number: MFCD02683863
CAS Number: 5471-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5752b356d03d342624f3aa51c64e27d9150aa8ff&vid=5752b356d03d342624f3aa51c64e27d9150aa8ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR