1-Isopropyl-4-piperidone ≥97.0% (by titrimetric analysis)
Supplier: TCI America
I0566-25G
I0566-5G
TCI0566-025GEA
84.7
USD
TCI0566-025G
76219-984
1-Isopropyl-4-piperidone ≥97.0% (by titrimetric analysis)
1-Isopropyl-4-piperidone
CAS Number: 5355-68-0
MDL Number: MFCD00038035
Molecular Formula: C8H15NO
Molecular Weight: 141.21
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 101
Specific Gravity (20/20): 0.95
Formula:
C₈H₁₅NO Boiling Pt: 101 °C Density: 0.95 (20 °C) |
MDL Number:
MFCD00038035 CAS Number: 5355-68-0 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...