Isonicotinic acid-N-oxide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Isonicotinic acid N-oxide, 1-Oxidopyridin-1-ium-4-carboxylic acid, 4-Carboxypyridine 1-oxide, Pyridine-4-carboxylic acid N-oxide

I0500-25G I0500-100G
TCI0500-025GEA 136 USD
TCI0500-025G TCI0500-100G
Isonicotinic acid-N-oxide ≥98.0% (by GC, titration analysis)
Isonicotinic acid N-oxide

CAS Number: 13602-12-5
MDL Number: MFCD00006209
Molecular Formula: C6H5NO3
Molecular Weight: 139.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 270


Other Product Names / Synonyms: Pyridine-4-carboxylic Acid N-Oxide

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 270 °C
MDL Number: MFCD00006209
CAS Number: 13602-12-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c12142656fc7ef674d4baf69af431bacfc341821&vid=c12142656fc7ef674d4baf69af431bacfc341821&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR