Tetrabutylammonium dihydrogen phosphate 0.5 mol/L in water
Supplier: TCI America
I0367-10ML
I0367-100ML
TCI0367-010MLEA
22.78
USD
TCI0367-010ML
TCI0367-100ML
Tetrabutylammonium dihydrogen phosphate 0.5 mol/L in water
Tetrabutylammonium dihydrogen phosphate
[Reagent for Ion-Pair Chromatography]
CAS Number: 5574-97-0
MDL Number: MFCD00064526
Molecular Formula: C16H37NO
Form: Clear Liquid
Color: Colorless
Other Product Names / Synonyms: IPC-TBA-P
Formula:
C₁₆H₃₈NO₄P |
MDL Number:
MFCD00064526 CAS Number: 5574-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...