(R)-(-)-1-(1-Naphthyl)ethyl isocyanate ≥90.0%
Supplier: TCI America
CAS Number: 42340-98-7
MDL Number: MFCD00004015
Molecular Formula: C13H11NO
Molecular Weight: 197.24
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Flash Point (°C): 93
Specific Gravity (20/20): 1.13
Specific rotation [a]20/D: -45 deg (C=5, Toluene)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. Determination (HPLC Labeling Reagents), e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: Isocyanic Acid (R)-(-)-1-(1-Naphthyl)ethyl Ester
Formula:
C₁₃H₁₁NO MW: 197.24 g/mol Density: 1.13 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD00004015 CAS Number: 42340-98-7 |
Specification Test Results
Purity/Analysis Method | >90.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...