DL-Isoleucine ≥98.0%
Supplier: TCI America
Synonyms:
H-DL-Ile-OH
I0180-5G
I0180-25G
TCI0180-5GEA
40
USD
TCI0180-5G
TCI0180-25G
DL-Isoleucine ≥98.0%
DL-Isoleucine
CAS Number: 443-79-8
MDL Number: MFCD00004268
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Ile-OH
Formula:
C₆H₁₃NO₂ MW: 131.17 g/mol Melting Pt: 275 |
MDL Number:
MFCD00004268 CAS Number: 443-79-8 Merck Index: 13,05198 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...