4-Nitrophenyl isocyanate ≥98.0% (contains varying amounts of polymers)
Supplier: TCI America
Synonyms:
1-Isocyanato-4-nitrobenzene
I0129-25G
I0129-5G
TCI0129-25GEA
295
USD
TCI0129-25G
TCI0129-5G
4-Nitrophenyl isocyanate ≥98.0% (contains varying amounts of polymers)
4-Nitrophenyl isocyanate
CAS Number: 100-28-7
MDL Number: MFCD00007306
Molecular Formula: C7H4N2O3
Molecular Weight: 164.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 162
Melting point (°C): 58
Flash Point (°C): 110
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Isocyanic Acid 4-Nitrophenyl Ester
Formula:
C₇H₄N₂O₃ MW: 164.12 g/mol Boiling Pt: 162 °C Melting Pt: 58 °C Flash Pt: 110 °C |
MDL Number:
MFCD00007306 CAS Number: 100-28-7 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...