3-(4-Hydroxy-3-methoxyphenyl)propionic acid ≥98.0%
Supplier: TCI America
Synonyms:
Dihydroferulic acid, 3-(4-Hydroxy-3-methoxyphenyl)propanoic acid
H1180-5G
H1180-1G
TCH1180-5GEA
200
USD
TCH1180-5G
TCH1180-1G
3-(4-Hydroxy-3-methoxyphenyl)propionic acid ≥98.0%
3-(4-Hydroxy-3-methoxyphenyl)propionic acid
CAS Number: 1135-23-5
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 91
Formula:
C₁₀H₁₂O₄ Melting Pt: 91 °C |
MDL Number:
MFCD00016558 CAS Number: 1135-23-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...