1-Hexyl-3-methylimidazolium tetrafluoroborate ≥97.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
1-n-Hexyl-3-methylimidazolium tetrafluoroborate, [HMIM] [BF4]
H1099-25G
H1099-5G
TCH1099-25GEA
155
USD
TCH1099-25G
TCH1099-5G
1-Hexyl-3-methylimidazolium tetrafluoroborate ≥97.0% (by total nitrogen basis)
1-Hexyl-3-methylimidazolium tetrafluoroborate
CAS Number: 244193-50-8
Molecular Formula: C10H19BF4N2
Molecular Weight: 254.08
Purity/Analysis Method:
Formula:
C₁₀H₁₉BF₄N₂ MW: 254.08 g/mol Density: 1.15 (20 °C) |
MDL Number:
MFCD03095459 CAS Number: 244193-50-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...