D-(-)-Histidine ≥99.0%
Supplier: TCI America
Synonyms:
(R)-(-)-Histidine, D-3-Imidazol-4-ylalanine, (2R)-(-)-Histidine, H-D-His-OH
H0998-25G
H0998-5G
TCH0998-25GEA
151
USD
TCH0998-25G
TCH0998-5G
D-(-)-Histidine ≥99.0%
D-(-)-Histidine
CAS Number: 351-50-8
MDL Number: MFCD00065963
Molecular Formula: C6H9N3O2
Molecular Weight: 155.16
Purity/Analysis Method: >99.0% (T,HPLC)
Form: Crystal
Specific rotation [a]20/D: -13 deg (C=11, 6mol/L HCl)
Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-His-OH
Formula:
C₆H₉N₃O₂ MW: 155.16 g/mol Melting Pt: 287 |
MDL Number:
MFCD00065963 CAS Number: 351-50-8 Merck Index: 12,04758 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (T,HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...