(4-Hydroxyphenoxy)acetic acid

Supplier: TCI America

H0992-25G H0992-5G
TCH0992-25GEA 261 USD
TCH0992-25G TCH0992-5G
(4-Hydroxyphenoxy)acetic acid
(4-Hydroxyphenoxy)acetic acid

CAS Number: 1878-84-8
MDL Number: MFCD00014362
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Form: Crystal
Color: White
Melting point (°C): 155

Formula: (C₆H₄(OH))OCH₂COOH
MW: 168.15 g/mol
MDL Number: MFCD00014362
CAS Number: 1878-84-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27ed5929a7ba946ca9057787fed3c4af31b97e19&vid=27ed5929a7ba946ca9057787fed3c4af31b97e19&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR