8-Hydroxyjulolidine ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1,2,3,5,6,7-Hexahydropyrido[3,2,1-ij]quinolin-8-ol, 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol

H0928-1G H0928-5G
TCH0928-001GEA 56 USD
TCH0928-001G TCH0928-005G
8-Hydroxyjulolidine ≥97.0% (by HPLC, titration analysis)
8-Hydroxyjulolidine

CAS Number: 41175-50-2
MDL Number: MFCD00006918
Molecular Formula: C12H15NO
Molecular Weight: 189.26
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 132


Other Product Names / Synonyms: 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol

Formula: C₁₂H₁₅NO
MW: 189.26 g/mol
Melting Pt: 132 °C
MDL Number: MFCD00006918
CAS Number: 41175-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7f5c81241122fd375107634ba6a9fe567f1a956&vid=a7f5c81241122fd375107634ba6a9fe567f1a956&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR