2-Hydroxy-6-methylnicotinic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-Hydroxy-6-methylpyridine-3-carboxylic acid
H0757-5G
H0757-25G
TCH0757-5GEA
44
USD
TCH0757-5G
TCH0757-025G
2-Hydroxy-6-methylnicotinic acid ≥98.0% (by titrimetric analysis)
2-Hydroxy-6-methylnicotinic acid
CAS Number: 38116-61-9
MDL Number: MFCD00006272
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 2-Hydroxy-6-methylpyridine-3-carboxylic Acid
Formula:
C₇H₇NO₃ MW: 153.14 g/mol |
MDL Number:
MFCD00006272 CAS Number: 38116-61-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...