4-(4-Hydroxyphenyl)benzonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4'-Hydroxy-4-biphenylcarbonitrile, 4'-Hydroxybiphenyl-4-carbonitrile, 4'-Hydroxy-[1,1'-biphenyl]-4-carbonitrile

H0523-25G H0523-5G
TCH0523-025GEA 140 USD
TCH0523-025G TCH0523-005G
4-(4-Hydroxyphenyl)benzonitrile ≥98.0% (by GC, titration analysis)
4-(4-Hydroxyphenyl)benzonitrile

CAS Number: 19812-93-2
MDL Number: MFCD00059625
Molecular Formula: C13H9NO
Molecular Weight: 195.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 198


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4'-Hydroxy-4-biphenylcarbonitrile

Formula: C₁₃H₉NO
MW: 195.22 g/mol
Melting Pt: 198 °C
MDL Number: MFCD00059625
CAS Number: 19812-93-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65168a3dea96701e636bb055bae860055997ce2e&vid=65168a3dea96701e636bb055bae860055997ce2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR