2-Hexyldecanoic acid ≥98.0%

Supplier: TCI America

Synonyms: Isopalmitic Acid, Pentadecane-7-carboxylic Acid

H0507-25ML H0507-500ML H0507-100ML
TCH0507-025MLEA 25.44 USD
TCH0507-025ML TCH0507-500ML TCH0507-100ML
2-Hexyldecanoic acid ≥98.0%
2-Hexyldecanoic acid

CAS Number: 25354-97-6
MDL Number: MFCD00059651
Molecular Formula: C16H32O2
Molecular Weight: 256.43
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 174
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Isopalmitic Acid, Pentadecane-7-carboxylic Acid

Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 150 °C
Density: 0.88 (20 °C)
Flash Pt: 174 °C
MDL Number: MFCD00059651
CAS Number: 25354-97-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6332d76d61ea1cbcf1226813ad6e5f3bdfc11ba&vid=b6332d76d61ea1cbcf1226813ad6e5f3bdfc11ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR