Ethylene glycol monohexyl ether ≥98.0%

Supplier: TCI America

Synonyms: Hexyl Cellosolve, 2-(Hexyloxy)ethanol

H0343-500ML H0343-25ML
TCH0343-500MLEA 175.01 USD
TCH0343-500ML TCH0343-25ML
Ethylene glycol monohexyl ether ≥98.0%
Ethylene glycol monohexyl ether

CAS Number: 112-25-4
MDL Number: MFCD00045997
Molecular Formula: C8H18O2
Molecular Weight: 146.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Melting point (°C): -45
Flash Point (°C): 102
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Hexyl Cellosolve, 2-(Hexyloxy)ethanol

Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 205 °C
Melting Pt: –45 °C
Density: 0.89 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00045997
CAS Number: 112-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51fd3679e01c28626854d19eaa51f1a649de1357&vid=51fd3679e01c28626854d19eaa51f1a649de1357&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR