2-Hydrazinoquinoline ≥98.0%

Supplier: TCI America

Synonyms: 2-Quinolylhydrazine, 2-Hydrazinylquinoline

H0178-25G H0178-5G H0178-1G
TCH0178-25GEA 938.4 USD
TCH0178-25G TCH0178-5G TCH0178-001G
2-Hydrazinoquinoline ≥98.0%
2-Hydrazinoquinoline

CAS Number: 15793-77-8
MDL Number: MFCD00041860
Molecular Formula: C9H9N3
Molecular Weight: 159.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 142


Other Product Names / Synonyms: 2-Quinolylhydrazine

Formula: C₉H₉N₃
MW: 159.19 g/mol
Melting Pt: 142 °C
MDL Number: MFCD00041860
CAS Number: 15793-77-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1af14b7efbfd2ad1accdb2690166e581ce6a58ca&vid=1af14b7efbfd2ad1accdb2690166e581ce6a58ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR