Gallium(III) chloride, anhydrous ≥98%
Supplier: TCI America
Synonyms:
Gallium trichloride
G0359-25G
G0359-5G
TCG0359-25GEA
162.11
USD
TCG0359-25G
TCG0359-5G
Gallium(III) chloride, anhydrous ≥98%
Gallium(III) chloride
CAS Number: 13450-90-3
MDL Number: MFCD00011018
Molecular Formula: Cl3Ga
Molecular Weight: 176.07
Form: Crystal
Color: White
Boiling point (°C): 201
Melting point (°C): 78
Formula:
Cl₃Ga MW: 176.08 g/mol Boiling Pt: 225 Melting Pt: 77.9 |
MDL Number:
MFCD00011018 CAS Number: 13450-90-3 UN: 3260 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...