Glycocyamine ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Glycocyamin, N-Amidinoglycine, 2-Guanidinoacetic acid, Guanidinoacetic Acid, Guanidoacetic Acid
G0167-500G
G0167-25G
TCG0167-500GEA
385
USD
TCG0167-500G
TCG0167-025G
Glycocyamine ≥97.0% (by titrimetric analysis)
Glycocyamine
CAS Number: 352-97-6
MDL Number: MFCD00004278
Molecular Formula: C3H7N3O2
Molecular Weight: 117.11
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: Guanidoacetic Acid, Guanidinoacetic Acid
Formula:
C₃H₇N₃O₂ MW: 117.11 g/mol |
MDL Number:
MFCD00004278 CAS Number: 352-97-6 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...