5-Fluorooxindole ≥98.0%
Supplier: TCI America
Synonyms:
5-Fluoroindolin-2-one, 5-Fluoro-2-oxoindoline, 5-Fluoro-2-oxindole
F0699-1G
F0699-5G
TCF0699-1GEA
60.7
USD
TCF0699-1G
TCF0699-5G
5-Fluorooxindole ≥98.0%
5-Fluorooxindole
CAS Number: 56341-41-4
MDL Number: MFCD02179598
Molecular Formula: C8H6FNO
Molecular Weight: 151.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 145
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Fluoro-2-indolinone
Formula:
C₈H₆FNO MW: 151.14 g/mol Melting Pt: 145 °C |
MDL Number:
MFCD02179598 CAS Number: 56341-41-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...