4-Fluorosalicylic acid ≥98.0%
Supplier: TCI America
Synonyms:
4-Fluoro-2-hydroxybenzoic acid
F0637-1G
F0637-25G
F0637-5G
TCF0637-1GEA
47
USD
TCF0637-1G
TCF0637-25G
TCF0637-5G
4-Fluorosalicylic acid ≥98.0%
4-Fluorosalicylic acid
CAS Number: 345-29-9
MDL Number: MFCD00153168
Molecular Formula: C7H5FO3
Molecular Weight: 156.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 189
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-2-hydroxybenzoic Acid
Formula:
C₇H₅FO₃ Melting Pt: 189 °C |
MDL Number:
MFCD00153168 CAS Number: 345-29-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...