2-Fluoro-6-nitrophenol ≥98.0%
Supplier: TCI America
F0623-5G
F0623-1G
TCF0623-5GEA
96
USD
TCF0623-5G
TCF0623-1G
2-Fluoro-6-nitrophenol ≥98.0%
2-Fluoro-6-nitrophenol
CAS Number: 1526-17-6
MDL Number: MFCD00042446
Molecular Formula: C6H4FNO3
Molecular Weight: 157.10
Purity/Analysis Method:
Formula:
C₆H₄FNO₃ Melting Pt: 92 °C |
MDL Number:
MFCD00042446 CAS Number: 1526-17-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...