N-Fmoc-D-leucine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Fmoc-D-Leu-OH
F0603-5G
F0603-1G
TCF0603-5GEA
45
USD
TCF0603-5G
TCF0603-1G
N-Fmoc-D-leucine ≥98.0% (by HPLC, titration analysis)
N-Fmoc-D-leucine
CAS Number: 114360-54-2
MDL Number: MFCD00062957
Molecular Formula: C21H23NO4
Molecular Weight: 353.42
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 155
Specific rotation [a]20/D: 25 deg (C=1, DMF)
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-D-leucine, Fmoc-D-Leu-OH
Formula:
C₂₁H₂₃NO₄ MW: 353.4 g/mol Melting Pt: 155 °C |
MDL Number:
MFCD00062957 CAS Number: 114360-54-2 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...