4-Fluoro-m-toluidine ≥98.0%
Supplier: TCI America
Synonyms:
4-Fluoro-3-methylaniline, 5-Amino-2-fluorotoluene
F0527-25G
F0527-5G
TCF0527-25GEA
57
USD
TCF0527-25G
TCF0527-5G
4-Fluoro-m-toluidine ≥98.0%
4-Fluoro-m-toluidine
CAS Number: 452-69-7
Molecular Formula: C7H8FN
Molecular Weight: 125.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 86
Melting point (°C): 36
Flash Point (°C): 104
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Amino-2-fluorotoluene, 4-Fluoro-m-toluidine
Formula:
C₇H₈FN MW: 125.15 Boiling Pt: 86 °C Melting Pt: 36 Density: 1.12 Flash Pt: 104 °C |
MDL Number:
MFCD00025294 CAS Number: 452-69-7 UN: 2811 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...