5-Fluorosalicylic acid ≥98.0%
Supplier: TCI America
Synonyms:
5-Fluoro-2-hydroxybenzoic acid
F0469-5G
F0469-1G
TCF0469-5GEA
137
USD
TCF0469-5G
TCF0469-1G
5-Fluorosalicylic acid ≥98.0%
5-Fluorosalicylic acid
CAS Number: 345-16-4
MDL Number: MFCD00002456
Molecular Formula: C7H5FO3
Molecular Weight: 156.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 180
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Fluoro-2-hydroxybenzoic Acid, 2-Hydroxy-5-fluorobenzoic Acid
Formula:
C₇H₅FO₃ Melting Pt: 180 °C |
MDL Number:
MFCD00002456 CAS Number: 345-16-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...