N(α)-Fmoc-L-tryptophan ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Fmoc-Trp-OH
F0307-5G
F0307-25G
TCF0307-005GEA
33
USD
TCF0307-005G
TCF0307-25G
N(α)-Fmoc-L-tryptophan ≥98.0% (by HPLC, titration analysis)
N(α)-Fmoc-L-tryptophan
CAS Number: 35737-15-6
MDL Number: MFCD00037126
Molecular Formula: C26H22N2O4
Molecular Weight: 426.47
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 187
Specific rotation [a]20/D: -28.5 deg (C=1, DMF)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-L-tryptophan, Fmoc-Trp-OH
Formula:
C₂₆H₂₂N₂O₄ MW: 426.5 g/mol Melting Pt: 187 °C |
MDL Number:
MFCD00037126 CAS Number: 35737-15-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...