Fmoc-L-phenylalanine ≥98.0%

Supplier: TCI America

Synonyms: N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine, N-α-Fmoc-L-Phenylalanine, Fmoc-Phe-OH, N-Fmoc-L-phenylalanine

F0297-25G F0297-5G
TCF0297-25GEA 75 USD
TCF0297-25G TCF0297-005G
Fmoc-L-phenylalanine ≥98.0%
Fmoc-L-phenylalanine

CAS Number: 35661-40-6
MDL Number: MFCD00037128
Molecular Formula: C24H21NO4
Molecular Weight: 387.44
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 183
Specific rotation [a]20/D: -39.5 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-L-phenylalanine, Fmoc-Phe-OH

Formula: C₂₄H₂₁NO₄
MW: 387.4 g/mol
Melting Pt: 183 °C
MDL Number: MFCD00037128
CAS Number: 35661-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0774c35fd592aaeb54f558b2766aec87ad65851d&vid=0774c35fd592aaeb54f558b2766aec87ad65851d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR