N-Fmoc-L-leucine
Supplier: TCI America
F0295-25G
F0295-1G
F0295-5G
TCF0295-25GEA
71.1
USD
TCF0295-25G
TCF0295-001G
TCF0295-005G
N-Fmoc-L-leucine
N-Fmoc-L-leucine
CAS Number: 35661-60-0
MDL Number: MFCD00037133
Molecular Formula: C21H23NO4
Molecular Weight: 353.42
Form: Crystal
Color: White
Melting point (°C): 154
Specific rotation [a]20/D: -25 deg (C=1, DMF)
Other Product Names / Synonyms: N-Fmoc-L-leucine, Fmoc-Leu-OH
Formula:
C₂₁H₂₃NO₄ MW: 353.4 g/mol Melting Pt: 154 °C |
MDL Number:
MFCD00037133 CAS Number: 35661-60-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...