N-Fmoc-glycine ≥98.0%
Supplier: TCI America
Synonyms:
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetic acid, Fmoc-Gly-OH
F0293-25G
F0293-5G
TCF0293-25GEA
42
USD
TCF0293-25G
TCF0293-005G
N-Fmoc-glycine ≥98.0%
N-Fmoc-glycine
CAS Number: 29022-11-5
MDL Number: MFCD00037140
Molecular Formula: C17H15NO4
Molecular Weight: 297.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 176
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-glycine, Fmoc-Gly-OH
Formula:
C₁₇H₁₅NO₄ MW: 297.3 g/mol Melting Pt: 176 °C |
MDL Number:
MFCD00037140 CAS Number: 29022-11-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...