Flavanone ≥98.0%

Supplier: TCI America

Synonyms: 2-Phenylchroman-4-one

F0255-25G F0255-5G
TCF0255-025GEA 64 USD
TCF0255-025G TCF0255-5G
Flavanone ≥98.0%
Flavanone

CAS Number: 487-26-3
MDL Number: MFCD00006841
Molecular Formula: C15H12O2
Molecular Weight: 224.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 76


Product Categories: Bioscience, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 2,3-Dihydro-2-phenyl-4H-benzopyran-4-one, 2-Phenylchromanone

Formula: C₁₅H₁₂O₂
MW: 224.26
Melting Pt: 76 °C
MDL Number: MFCD00006841
CAS Number: 487-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5181aeebd94796177f7e1a3c12b437ccf5c05146&vid=5181aeebd94796177f7e1a3c12b437ccf5c05146&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR