Aminoiminomethanesulfinic acid ≥93.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Thiourea dioxide, FormamidinesulphInic acid
F0115-500G
F0115-25G
TCF0115-500GEA
204
USD
TCF0115-500G
TCF0115-25G
Aminoiminomethanesulfinic acid ≥93.0% (by titrimetric analysis)
Aminoiminomethanesulfinic acid
CAS Number: 1758-73-2
MDL Number: MFCD00002397
Molecular Formula: CH4N2O2S
Molecular Weight: 108.12
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Melting point (°C): 126
Other Product Names / Synonyms: Thiourea dioxide
Formula:
CH₄N₂O₂S MW: 108.12 g/mol Melting Pt: 126 °C |
MDL Number:
MFCD00002397 CAS Number: 1758-73-2 UN: 3341 ADR: 4.2,III |
|
Specification Test Results
Purity/Analysis Method | >93.0% (T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...