Formamidine disulfide dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: Dithiourea Dihydrochloride, Dithiodiformamide Dihydrochloride

F0046-5G F0046-25G
TCF0046-5GEA 22 USD
TCF0046-5G TCF0046-025G
Formamidine disulfide dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
Formamidine disulfide dihydrochloride

CAS Number: 14807-75-1
MDL Number: MFCD00012586
Molecular Formula: C2H6N4S2
Molecular Weight: 223.13
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal


Other Product Names / Synonyms: Dithiodiformamide Dihydrochloride, Dithiourea Dihydrochloride

Formula: C₂H₆N₄S₂
MW: 223.13 g/mol
MDL Number: MFCD00012586
CAS Number: 14807-75-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f63637d96d83e28bc60b37af96ee30d67b80cafc&vid=f63637d96d83e28bc60b37af96ee30d67b80cafc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR