Ethyl-4,4,4-trifluorobutyrate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 4,4,4-trifluorobutanoate, Ethyl 4,4,4-trifluorobutyrate

E0830-1G E0830-5G
TCE0830-1GEA 21 USD
TCE0830-1G TCE0830-5G
Ethyl-4,4,4-trifluorobutyrate ≥98.0%
Ethyl-4,4,4-trifluorobutyrate

CAS Number: 371-26-6
MDL Number: MFCD00041398
Molecular Formula: C6H9F3O2
Molecular Weight: 170.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Flash Point (°C): 33
Specific Gravity (20/20): 1.16


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 4,4,4-Trifluorobutyric Acid Ethyl Ester

Formula: C₆H₉F₃O₂
MW: 170.13 g/mol
Boiling Pt: 127 °C
Density: 1.16 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00041398
CAS Number: 371-26-6
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b45ab167ba2cda2624d68ba9637d44f908c2a89&vid=1b45ab167ba2cda2624d68ba9637d44f908c2a89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR