5-Ethyluracil ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Ethyluracil, 5-Ethylpyrimidine-2,4(1H,3H)-dione

E0807-1G E0807-5G
TCE0807-1GEA 24 USD
TCE0807-1G TCE0807-5G
5-Ethyluracil ≥98.0% (by HPLC, titration analysis)
5-Ethyluracil

CAS Number: 4212-49-1
MDL Number: MFCD00079187
Molecular Formula: C6H8N2O2
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 303
Lambda max.: 265 nm (EtOH)


Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 2,4-Dihydroxy-5-ethylpyrimidine

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Melting Pt: 303 °C
MDL Number: MFCD00079187
CAS Number: 4212-49-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d271ad709e2e2123653fd7b2b7c9ab46ccc7c719&vid=d271ad709e2e2123653fd7b2b7c9ab46ccc7c719&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR