Ethyl-2-methylvalerate ≥98.0%

Supplier: TCI America

Synonyms: 2-Methylpentanoic Acid Ethyl Ester, Ethyl 2-methylpentanoate, 2-Methylvaleric Acid Ethyl Ester, Ethyl 2-Methylpentanoate

E0801-25ML E0801-500ML
TCE0801-25MLEA 25 USD
TCE0801-25ML TCE0801-500ML
Ethyl-2-methylvalerate ≥98.0%
Ethyl-2-methylvalerate

CAS Number: 39255-32-8
MDL Number: MFCD00036564
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 39
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 2-Methylvaleric Acid Ethyl Ester, Ethyl 2-Methylpentanoate, 2-Methylpentanoic Acid Ethyl Ester

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 153 °C
Density: 0.87 (20 °C)
Flash Pt: 39 °C
MDL Number: MFCD00036564
CAS Number: 39255-32-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfce419400698cdbb329f0eb9572dacbf295de4f&vid=bfce419400698cdbb329f0eb9572dacbf295de4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR